Learn More
Cayman Chemical 3 4MethylendIoxyNbenzylca 10mg
SDP
Supplier:  Cayman Chemical 900133010 MG
Encompass_Preferred
A substituted cathinone that is structurally analogous to methylone, a narcotic which is regulated in the United States; intended for research and forensic applications
Catalog No. 50-289-4198