Learn More
Medchemexpress LLC 3 5 3 -Triiodo-D-thy 10mg | 1393257 | 650.97 g/mol | C15H12I3NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11632510MG
Encompass_Preferred
3,5,3'-Triiodo-D-thyronine is a research-grade stereoisomer of the thyroid hormone triiodothyronine (T3), supplied as a purified solid for biochemical and pharmacological studies. It is reported to be less effective than the alternative T isomer in normalizing elevated plasma creatine phosphokinase (CPK) activity and is used in in vitro assays and mechanistic investigations of thyroid hormone action.
  • High purity suitable for laboratory research.
  • Available in small quantities for dose-response and pilot studies.
  • Supplied as a stable solid for standard storage and handling.
  • Applicable to metabolic and endocrine mechanism studies.
Catalog No. 50-002-26578