Learn More
Medchemexpress LLC 3 5 3 -Triiodo-D-thy 5mg | 1393257 | 650.97 g/mol | C15H12I3NO4 | 5 MG

Supplier: Medchemexpress LLC HY1163255MG
3,5,3'-Triiodo-D-thyronine is the D-isomer of the thyroid hormone triiodothyronine (T3), supplied as a solid research reagent for biochemical and pharmacological studies; supplier notes it is less effective than the corresponding T isomer in normalizing increased plasma creatine phosphokinase (CPK) activity.
- Research reagent in solid powder form
- Molecular formula C15H12I3NO4; molecular weight 650.97 g/mol
- Purity 95.3% (as supplied)
- Suitable for in vitro biochemical and pharmacological assays
- Available in small laboratory quantities for research use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.