Learn More
Medchemexpress LLC (3α,5β,6β,7β)-6-ethyl-cholane-3,7,24-triol | 1632118-69-4 | MFCD30738007 | 695.0% | 406.64 g·mol⁻1 | C26H46O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10127410MM 1ML
Encompass_Preferred
BAR501 is a synthetic small-molecule agonist of the G protein-coupled bile acid receptor GPBAR1 (TGR5). It selectively activates GPBAR1 with an EC50 of about 1 μM in HEK293 CRE-luciferase transactivation assays and is supplied for research applications in solution or solid form.
  • Provides selective GPBAR1 agonism with an EC50 ≈ 1 μM.
  • Transactivates GPBAR1 in HEK293 cells as measured by CRE-driven luciferase reporter assays.
  • Available as a 10 mM solution in DMSO (1 mL) or as solid vials for reconstitution.
  • Molecular formula C26H46O3 and molecular weight ~406.64 g·mol⁻1.
  • High purity suitable for biological assays (reported ≥95%).
  • Intended for research use only; not for human or veterinary applications.
Catalog No. 50-002-11714