Learn More
Medchemexpress LLC 3-(5,6-dihydro-4H-pyrrolo[3,2,1-ij]quinolin-1-yl)-4-(1H-indol-3-yl)pyrrolidine-2,5-dione | 1239986-50-5 | MFCD18917093 | 95.2% | 369.42 | C23H19N3O2 | 10 MG

Supplier: Medchemexpress LLC HY7668810MG
(Rac)-Tivantinib is the racemic form of Tivantinib, a selective c-Met tyrosine kinase inhibitor provided for use as an experimental control in biochemical and cellular research. Supplied as a powder with recommended low-temperature storage, it has CAS 1239986-50-5 and a molecular weight of 369.42 g·mol⁻1.
- Racemic isomer suitable as an experimental control in c-Met inhibition studies.
- Powder form allows accurate weighing and formulation of stock solutions.
- Purity typically reported near 95% for research-grade applications.
- Stable under recommended low-temperature storage for extended shelf life.
- Soluble in common organic solvents for preparation of assay concentrations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.