Learn More
Medchemexpress LLC 3,5-bis[(4-methylphenyl)methylene]-4-piperidone hydrochloride | 157654-67-6 | MFCD09753281 | 98.9% | 339.86 g/mol | C21H22ClNO | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10070850MG
Encompass_Preferred
NSC632839 is a small-molecule, nonselective isopeptidase inhibitor used in biochemical and cell-based research to probe deubiquitinating enzymes and SUMO proteases. It shows mid-micromolar activity against USP2 and USP7 and greater potency against SENP2, and is supplied as a high-purity solid suitable for mechanistic studies.
  • Nonselective isopeptidase inhibitor active against USP2, USP7, and SENP2 (EC50s: 45±4 μM, 37±1 μM, 9.8±1.8 μM).
  • High purity suitable for research (≈98.9%).
  • Solid form, light yellow to yellow appearance for easy handling.
  • Soluble in DMSO (~4 mg/mL) with warming or sonication if required.
  • Stable when stored sealed away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-14536