Learn More
Medchemexpress LLC 3,5-bis(trifluoromethyl)benzyl azabicyclo[3.1.0]hexane carboxylate | 2162961-71-7 | ≥98.0% | 527.4 | C23H19F6N5O3 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12477225MG
Encompass_Preferred
BI-2545 is a small-molecule chemical probe developed for research that potently inhibits autotaxin (ATX) and is suitable for both in vitro and in vivo studies. It demonstrates single-digit nanomolar biochemical potency against ATX and has been shown to lower plasma lysophosphatidic acid (LPA) levels following oral dosing in rodents.
Catalog No. 50-002-35851