Learn More
Medchemexpress LLC 3-[5-bromo-6-[(2R,3R)-2-(4-fluorophenyl)-3-(hydroxymethyl)morpholine-4-carbonyl]-1-methylbenzimidazol | 2259620-80-7 | 98.9% | 616.48 g/mol | C31H27BrFN5O3 | 1 ML

Supplier: Medchemexpress LLC HY11731810MM 1ML
PDE12-IN-1 is a potent, selective inhibitor of phosphodiesterase 12 supplied as a 10 mM solution in DMSO (1 mL). It increases 2′,5′-linked adenylate polymers (2-5A) and has reported antiviral activity. Intended for laboratory research use only; not for human or clinical applications.
- Potent PDE12 inhibition (pIC50 = 9.1).
- Increases 2-5A levels (pEC50 = 7.7).
- Supplied as a 10 mM solution in DMSO, 1 mL vial.
- High purity (98.85%).
- Molecular formula C31H27BrFN5O3; molecular weight 616.48 g/mol.
- Store sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
- For research use only; not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.