Learn More
Medchemexpress LLC 3,5-dibromo-4-methoxybenzoic acid | 4073-35-2 | MFCD00016511 | >98.0% | 309.94 g/mol | C8H6Br2O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0386271G
Encompass_Preferred
3,5-dibromo-4-methoxybenzoic acid is a brominated aromatic carboxylic acid used as a specialty research reagent and synthetic building block in organic chemistry and natural product studies. It is identified by CAS 4073-35-2, has molecular formula C8H6Br2O3, and a molecular weight of 309.94 g/mol.
  • Brominated aromatic carboxylic acid with defined substitution pattern
  • Useful building block for organic synthesis and derivatization
  • Characterized by CAS number 4073-35-2 and known molecular weight
  • Storage-stable under recommended conditions for extended shelf life
  • Available in research-scale quantities for laboratory use
Catalog No. 50-003-01673