Learn More
Accela Chembio Inc MF: C7H6Br2O | MW: 265.93 | >97% | Flash Point: 98.1 celsius | BP: 124 celsius at 760 mmHg.
Supplier:  Accela Chembio Inc SY01876325G
Encompass_Preferred
MF: C7H6Br2O | MW: 265.93 | >97% | Flash Point: 98.1 celsius | BP: 124 celsius at 760 mmHg.
Catalog No. 50-252-447