Learn More
Medchemexpress LLC 3,5-Dicaffeoyl-epi-quinic acid | 879305-14-3 | 97.7% | 516.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN98955MG
Encompass_Preferred
3,5-Dicaffeoyl-epi-quinic acid is a compound extracted from *Chrysanthemum morifolium*. It demonstrates significant antioxidant activities, effectively scavenging DPPH radical and superoxide anion radical with IC50s of 5.6 and 2.9 μg/mL, respectively.
  • Extracted from Chrysanthemum morifolium
  • Exhibits antioxidant activities
  • Scavenges DPPH radical and superoxide anion radical
  • IC50s of 5.6 and 2.9 μg/mL for DPPH and superoxide anion radicals
  • Appears as a white to off-white solid
Catalog No. 50-004-26365