Learn More
Medchemexpress LLC 3,5-Dicaffeoyl-epi-quinic acid | 879305-14-3 | 97.73% | 516.45 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN98951MG
Encompass_Preferred
3,5-Dicaffeoyl-epi-quinic acid is a compound that can be extracted from Chrysanthemum morifolium. It possesses antioxidant activities and is capable of scavenging DPPH radical and superoxide anion radical. Its IC50 values for scavenging DPPH radical and superoxide anion radical are 5.6 μg/mL and 2.9 μg/mL, respectively.
  • Extracted from Chrysanthemum morifolium.
  • Exhibits antioxidant activities.
  • Scavenges DPPH radical with an IC50 of 5.6 μg/mL.
  • Scavenges superoxide anion radical with an IC50 of 2.9 μg/mL.
Catalog No. 50-004-26736