Learn More
Medchemexpress LLC 3,5-dichloro-4-hydroxybenzonitrile | 1891-95-8 | MFCD00002177 | 188.01 g/mol | C7H3Cl2NO | 500 MG

Supplier: Medchemexpress LLC HYW059747500MG
Chloroxynil is a small-molecule research reagent (CAS 1891-95-8) used for analytical and biological studies. It has formula C7H3Cl2NO and a molecular weight of 188.01 g/mol. Solubility is reported as ≥ 5 mg/mL (26.59 mM) in certain solvents.
- Small-molecule research reagent for analytical use.
- CAS 1891-95-8; molecular formula C7H3Cl2NO.
- Molecular weight 188.01 g/mol.
- Reported solubility ≥ 5 mg/mL (26.59 mM) in recommended solvents.
- Pack-size availability varies by distributor; confirm specific package quantity before ordering.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.