Learn More
Medchemexpress LLC 3,5-dichloropyridine | 2457-47-8 | MFCD00006376 | 100.0% | 147.99 g/mol | C5H3Cl2N | 25 G

Supplier: Medchemexpress LLC 25GHYW015627
3,5-Dichloropyridine is a chlorinated pyridine derivative used as a chemical intermediate for organic synthesis. It is suitable for research applications that require a halogenated pyridine building block, including the synthesis of metal-organic framework (MOF) materials and P2X7 receptor antagonist compounds. The material is supplied as a high-purity reagent with supporting documentation for laboratory use.
- High purity (99.97%) suitable for research applications.
- Available in small to bulk pack sizes (25 g-250 g).
- Suitable as a building block for MOF synthesis and P2X7 receptor antagonist development.
- Provided with datasheet, COA, and SDS for traceability and safe handling.
- Molecular formula C5H3Cl2N; molecular weight 147.99 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.