Learn More
Medchemexpress LLC 3,5-Difluoropicolinonitrile | 298709-29-2 | 140.09 | 10 G

Supplier: Medchemexpress LLC HYW00808010G
3,5-Difluoropicolinonitrile is a high-purity chemical compound commonly appearing as a white to off-white solid. With a molecular weight of 140.09 and a purity of 99.8%, it is suitable for various laboratory research applications. This compound exhibits a melting point between 30-34°C and a boiling point of 187.8°C.
- Used as a biochemical reagent
- Suitable for laboratory research applications
- White to off-white solid appearance
- Molecular weight of 140.09
- Purity of 99.8% (GC)
- Melting point between 30-34°C
- Boiling point of 187.8°C
- Flash point of 90°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.