Learn More
Medchemexpress LLC 3,5-diiodothyropropionic acid | 1158-10-7 | MFCD00045945 | 99.2% | 510.06 g/mol | C15H12I2O4 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12623610MM 1ML
Encompass_Preferred
3,5-diiodothyropropionic acid is a thyroid hormone analog used to probe thyroid hormone receptor activity and metabolic effects in cellular and biochemical research. The compound is characterized by CAS 1158-10-7, molecular formula C15H12I2O4, and molecular weight 510.06 g/mol. It is supplied as a 10 mM solution (1 mL) and is also available in solid quantities for analytical or preparative use.
  • Useful for receptor binding and metabolic pathway studies.
  • Supplied as a ready-to-use 10 mM solution for in vitro assays.
  • Available in solid doses for storage or bulk use.
  • High purity suitable for analytical and research applications.
  • Characterized by CAS 1158-10-7 and established molecular data.
Catalog No. 50-002-35872