Learn More
Selleck Chemical LLC 3.5-Diiodotyrosine Dihydrate S3958-25mg
SDP
Supplier:  Selleck Chemical LLC S395825MG
Encompass_Preferred
3 5-Diiodotyrosine Dihydrate (Iodogorgoic acid apothyrin jodgorgon) is a product from the iodination of monoiodotyrosine In the biosynthesis of thyroid hormones diiodotyrosine residues are coupled with other monoiodotyrosine or diiodotyrosine residues to form T4 or T3 thyroid hormones
Catalog No. 50-310-8393