Learn More
Medchemexpress LLC 3,5-dimethoxybenzyl alcohol | 705-76-0 | MFCD00004641 | 99.9% | 168.19 | C9H12O3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017093
Encompass_Preferred
3,5-Dimethoxybenzyl alcohol is an aromatic benzyl alcohol used as a research chemical and synthetic intermediate in organic synthesis and natural-product studies. The compound has formula C9H12O3, molecular weight 168.19, and CAS 705-76-0. It is supplied as a solid with batch-specific analytical data and storage guidance for laboratory use.
  • High purity: 99.9% (HPLC).
  • White to off-white solid appearance.
  • Suitable as a synthetic intermediate and research reagent.
  • Batch-specific certificate of analysis available.
  • Storage guidance provided for ambient and frozen conditions.
Catalog No. 50-003-03825