Learn More
Medchemexpress LLC 3,5-Dimethoxyphenylboronic acid | 192182-54-0 | 99.6% | 181.98 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3292010G
Encompass_Preferred
3,5-Dimethoxyphenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is suitable for research purposes only and not for medical, clinical diagnostic, or food use in humans or animals.
  • Biochemical reagent for life science related research.
  • Suitable for use as a biological material or organic compound.
  • Can be used as a reactant for palladium-catalyzed coupling reactions.
  • Useful in the preparation of benzopyranone derivatives as positive GABAA receptor modulators.
  • Applicable for the preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling.
  • Acts as a reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide.
  • Used in the preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.
Catalog No. 50-004-03540