Learn More
Medchemexpress LLC 3,5-Dimethylbenzaldehyde | 5779-95-3 | 134.18 | 500 MG

Supplier: Medchemexpress LLC HYW002343500MG
3,5-Dimethylbenzaldehyde is a chemical compound used as a building block in chemical synthesis.
- Molecular formula: C9H10O
- Appearance: Liquid (density: 0.998 g/cm³)
- Color: Colorless to light yellow
- Initial source: Endogenous metabolite
- Structure classification: Ketones, aldehydes, acids
- Storage: 4°C, stored under nitrogen
- In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.