Learn More
Medchemexpress LLC 3,5-Heptanedione, 1,7-bis(4-hydroxy-3-methoxyphenyl)- | 36062-04-1 | 372.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN089310MM/1ML
Encompass_Preferred
Tetrahydrocurcumin is a Curcuminoid found in turmeric (Curcuma longa) that is produced by the reduction of Curcumin. It inhibits CYP2C9 and CYP3A4. It is for research use only and not sold to patients.
  • Produced by the reduction of Curcumin.
  • Inhibits CYP2C9 and CYP3A4.
  • For research use only.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • In solvent storage: -80°C for 2 years; -20°C for 1 year.
Catalog No. 50-004-16898