Learn More
CHEMSHUTTLE INC 3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)benzoic acid | 641571-18-8 | MFCD29477386 | 100mg
Supplier:  CHEMSHUTTLE INC 134595100MG
Encompass
3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)benzoic acid | Putity: 95% | MW: 270.21 | 641571-18-8 | MFCD29477386 | 100mg
Catalog No. NC1645521