Learn More
Medchemexpress LLC 3-(5-methyl-thiazol-2-yl)-5-[3R-oxolan-3-yl]oxy-N-[1R-1-(2-CF3-pyrimidin-5-yl)ethyl]benzamide | 1948229-21-7 | 99.9% | 478.5 g/mol | C22H21F3N4O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1091705MG
Encompass_Preferred
Eliapixant (BAY 1817080) is an investigational small-molecule antagonist of the P2X3 purinergic receptor. It has been studied in clinical and preclinical research for refractory chronic cough and other sensory neuropathic conditions. This compound is intended for laboratory research use only and is not approved for clinical use.
  • Potent P2X3 receptor antagonist with reported IC50 approximately 8 nM.
  • Useful for research on refractory chronic cough and sensory neuropathic conditions.
  • High reported purity (99.95%) suitable for analytical and preclinical studies.
  • Available in small research pack sizes for experimental workflows.
  • Molecular weight 478.5 g/mol; formula C22H21F3N4O3S.
Catalog No. 50-002-64571