Learn More
Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45-Pentadecaoxaheptatetracontan-1-ol | 6812-36-8 | 98.0% | C32H66O17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141235100MG
Encompass_Preferred
HO-PEG16-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds with two ligands connected by a linker: one for an E3 ubiquitin ligase and another for a target protein. They leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-56274