Learn More
Medchemexpress LLC 3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontan-1-amine, 35-azido- | 1800414-71-4 | ≥98.0% | C24H50N4O11 | 50 MG

Supplier: Medchemexpress LLC HY14021850MG
This Azido-PEG11-amine is a chemical compound with the molecular formula C24H50N4O11 and a molecular weight of 570.67. It appears as a colorless to light yellow liquid with a purity of at least 98.0%. This product is suitable for various laboratory applications.
- Colorless to light yellow liquid appearance
- Molecular weight of 570.67
- Purity of at least 98.0%
- Consistent with structure by 1H NMR Spectrum
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.