Learn More
Medchemexpress LLC 3,6,9,12,15,18,21,24-octaoxapentacosan-1-amine | 869718-81-0 | >98.0% | 383.48 | C17H37NO8 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW040236500MG
Encompass_Preferred
m-PEG8-Amine is a cleavable polyethylene glycol (PEG8) linker with a terminal primary amine used for conjugation in antibody-drug conjugates (ADCs) and other bioconjugation chemistries for research applications.
  • Cleavable ADC linker with PEG8 spacer and terminal primary amine for conjugation.
  • High purity: >98.0%.
  • Molecular weight 383.48 g/mol and formula C17H37NO8.
  • Suitable for ADC and PROTAC linker synthesis and bioconjugation research.
  • Available as 500 MG pack for laboratory use.
Catalog No. 50-002-04090