Learn More
Medchemexpress LLC 3,6,9,12,15,18,21-Heptaoxatricosan-1-ol, 23-azido- | 352439-36-2 | ≥97.0% | C16H33N3O8 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1407991G
Encompass_Preferred
Azide-PEG8-alcohol is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent contains an Azide group, which facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc and SPAAC reactions
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54967