Learn More
Medchemexpress LLC 3,6,9,12,15,18-Hexaoxanonadecan-1-amine | 184357-46-8 | ≥97.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1304085G
Encompass_Preferred
m-PEG6-Amine is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It also functions as a cleavable ADC linker employed in the synthesis of antibody-drug conjugates (ADCs).
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Cleavable ADC linker
  • Employed in the synthesis of antibody-drug conjugates (ADCs)
  • PROTACs exploit the intracellular ubiquitin-proteasome system for selective protein degradation
  • ADCs comprise an antibody attached to a cytotoxin via an ADC linker
Catalog No. 50-003-40225