Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxanonadecanoic acid | 16142-03-3 | 99.9% | 310.34 | C13H26O8 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1205371G
Encompass_Preferred
m-PEG5-CH2COOH is a methoxy-terminated polyethylene glycol linker with a terminal acetic acid group, used as a non-cleavable linker in antibody-drug conjugate synthesis and as a PEG-based linker in PROTAC construction. It is a high-purity solid (≈99.9%) with molecular weight 310.34 and is readily soluble in DMSO under ultrasonic assistance; handle and store sealed to avoid moisture.
Catalog No. 50-002-02419