Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxatriacontan-1-ol | 3055-96-7 | MFCD00042660 | 98.0% | 450.65 | C24H50O7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW2501981ML
Encompass_Preferred
Hexaethylene glycol monododecyl ether is a nonionic surfactant and solubilizing reagent for laboratory research use. It is commonly used to solubilize hydrophobic compounds, as a mild detergent in biochemical assays, and to aid membrane protein handling. Supplied as a high-purity liquid for small-volume laboratory applications.
  • Nonionic surfactant suitable for solubilization of hydrophobic molecules.
  • High purity (98.0%) for reproducible experimental results.
  • Liquid form convenient for pipetting and small-volume workflows.
  • Compatible with many biological assay buffers and protocols.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-81675