Learn More
Medchemexpress LLC 3,6,9,12,15,18-hexaoxatriacontan-1-ol | 3055-96-7 | MFCD00042660 | 98.0% | 450.65 g·mol⁻¹ | C24H50O7 | 5 ML
SDP
Supplier:  Medchemexpress LLC HYW2501985ML
Encompass_Preferred
Hexaethylene glycol monododecyl ether is a nonionic surfactant and biochemical reagent used in life-science research for membrane protein solubilization, micelle formation, and detergent applications. It is supplied as a high-purity research-grade chemical and should be stored sealed and protected from moisture.
  • Nonionic surfactant suitable for membrane protein solubilization.
  • Promotes micelle formation for biochemical and biophysical assays.
  • High purity (≥98.0%) supports reproducible experimental results.
  • Compatible with aqueous buffers and common laboratory solvents.
  • Available in small-volume packaging for flexible laboratory use.
  • Store sealed and away from moisture; cold storage recommended for solutions.
Catalog No. 50-002-81532