Learn More
Medchemexpress LLC 3,6,9,12,15-pentaoxaheptadecan-1-ol, 17-amino- | 39160-70-8 | 99.9% | 281.35 | C12H27NO6 | 1 G

Supplier: Medchemexpress LLC HY1269421G
Amino-PEG6-alcohol is a six-unit polyethylene glycol (PEG) linker with terminal amino and alcohol functional groups used for bioconjugation. It is commonly applied as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker, offering improved solubility and spacer flexibility.
- Provides amino and alcohol functional groups for conjugation.
- Six-unit PEG spacer increases solubility and flexibility.
- High purity: 99.85% (HPLC).
- Molecular weight: 281.35 g/mol; formula C12H27NO6.
- Solubility: DMSO ≥ 100 mg/mL.
- Storage: pure form -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).
- Available in small bench-scale packages, including 1 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.