Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,12,15-pentaoxaheptadecan-1-ol, 17-amino | 39160-70-8 | 99.9% | 281.35 g/mol | C12H27NO6 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY126942100MG

Encompass_Preferred

Amino-PEG6-alcohol is a PEG-based bifunctional linker containing six ethylene glycol units with terminal amino and hydroxyl groups. It provides a hydrophilic, flexible spacer for conjugation chemistry, commonly used in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs).

  • Cas number: 39160-70-8
  • Molecular formula: C12H27NO6
  • Molecular weight: 281.35 g/mol
  • Purity: 99.85%
  • Appearance: colorless to light yellow liquid (density 1.083 ± 0.06 g/cm3)
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Applications: hydrophilic spacer for ADC and PROTAC synthesis

Catalog No. 50-002-02086


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.