Learn More
Medchemexpress LLC 3,6,9,12,15-pentaoxaheptadecan-1-ol, 17-amino- | 39160-70-8 | 99.9% | 281.35 g/mol | C12H27NO6 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY126942250MG
Encompass_Preferred
Amino-PEG6-alcohol is a PEG-based bifunctional linker containing a terminal amino group and a terminal alcohol. It is a non-cleavable 6-unit polyethylene glycol spacer used in conjugation chemistry for antibody-drug conjugates and proteolysis targeting chimeras. High purity and defined molecular properties make it suitable for synthesis and research applications.
  • Non-cleavable 6-unit PEG linker
  • Terminal amino and alcohol functional groups
  • Suitable for ADC and PROTAC synthesis
  • High purity suitable for chemical synthesis
  • Molecular weight 281.35 g/mol; formula C12H27NO6
Catalog No. 50-002-03475