Learn More
Medchemexpress LLC 3,6,9,12,15-Pentaoxaheptadecane, 1,17-diazido- | 356046-26-9 | 100.0% | 332.36 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY133392
Encompass_Preferred
Azido-PEG5-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Reacts with alkyne-containing molecules
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Reacts with molecules containing DBCO or BCN groups
Catalog No. 50-003-65767