Learn More
Medchemexpress LLC 3,6,9,12,15-Pentaoxahexadecanoic acid | 16024-66-1 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130478100MG
Encompass_Preferred
m-PEG4-CH2COOH is a PEG-based PROTAC linker, a colorless to light yellow liquid with a density of 1.125 g/cm³. With a molecular weight of 266.29 and chemical formula C11H22O7, it is primarily utilized in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system by linking an E3 ubiquitin ligase to a target protein, thereby selectively degrading the target.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Appears as a colorless to light yellow liquid
  • Has a chemical formula of C11H22O7
  • Features a molecular weight of 266.29
Catalog No. 50-003-38646