Learn More
Medchemexpress LLC 3,6,9,12-tetraoxapentadec-14-yn-1-ol, 1-(4-methylbenzenesulfonate) | 875770-32-4 | 99.3% | 386.46 | C18H26O7S | 250 MG
SDP
Supplier:  Medchemexpress LLC HY130387250MG
Encompass_Preferred
Propargyl-PEG4-Tos is a PEG-based linker used as an alkyne click-chemistry reagent and cleavable linker for PROTAC and antibody-drug conjugate synthesis. It provides a hydrophilic PEG4 spacer to improve solubility and enable CuAAC functionalization.
  • Click chemistry alkyne for copper-catalyzed azide-alkyne cycloaddition
  • Cleavable ADC and PROTAC linker functionality
  • PEG4 spacer increases hydrophilicity and solubility
  • Confirmed identity by 1H NMR and LCMS
  • High purity (HPLC 99.3%)
Catalog No. 50-002-04026