Learn More
Medchemexpress LLC 3,6,9,12-Tetraoxatetradecan-1-ol, 14,14,14-trifluoro-, 1-(4-methylbenzenesulfonate) | 1872433-61-8 | MFCD29079383 | ≥97.0% | C17H25F3O7S | 50 MG

Supplier: Medchemexpress LLC HY14037950MG
This chemical product is 3,6,9,12-Tetraoxatetradecan-1-ol, 14,14,14-trifluoro-, 1-(4-methylbenzenesulfonate), provided as a colorless to light yellow viscous liquid. It has a molecular formula of C17H25F3O7S and a molecular weight of 430.44. The product is suitable for various laboratory applications requiring high purity.
- Purity of ≥97.0% (NMR)
- Appearance as a colorless to light yellow viscous liquid
- Consistent 1H NMR spectrum
- Molecular formula: C17H25F3O7S
- Molecular weight: 430.44
- Storage in pure form at -20°C for 3 years or 4°C for 2 years
- Storage in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.