Learn More
Medchemexpress LLC 3,6,9,12-Tetraoxatetradecan-1-ol, 14-bromo- | 957205-14-0 | 98.0% | C10H21BrO5 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1412141G
Encompass_Preferred
Bromo-PEG5-alcohol is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs work by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53190