Learn More
Medchemexpress LLC 3,6,9,12-Tetraoxatetradecan-1-ol, 14-bromo- | 957205-14-0 | 98.0% | C10H21BrO5 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141214
Encompass_Preferred
Bromo-PEG5-alcohol is a PEG-based PROTAC linker, instrumental in the synthesis of PROTACs. These specialized molecules leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins, offering a novel approach in therapeutic development.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-52566