Learn More
Medchemexpress LLC 3,6,9,15,18,21-Hexaoxa-12-azatricosane-1,23-diol | 63721-14-2 | 98.0% | C16H35NO8 | 50MG
SDP
Supplier:  Medchemexpress LLC HY14054850MG
Encompass_Preferred
NH-bis-PEG4 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for a target protein. They leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57028