Learn More
Medchemexpress LLC 3,6,9,16-Tetraoxaheptadecanoic acid, 17-phenyl-, 1,1-dimethylethyl ester | 2381196-81-0 | 95.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130619500MG
Encompass_Preferred
Boc-C1-PEG3-C4-OBn (PROTAC Linker 15) is a PROTAC linker characterized by its PEG composition. It can be utilized in the synthesis of various PROTACs, such as PROTAC SGK3 degrader-1 (HY-125878). PROTACs consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Characterized by PEG composition
  • Used in the synthesis of various PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40798