Learn More
Medchemexpress LLC 3,6,9-trioxa-12-azaheptadecan-17-oic acid, 1-amino-16-[[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)m | 710323-40-3 | 99.7% | 615.64 | C27H37N9O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1384845MG
Encompass_Preferred
Folate-PEG3-amine is a cleavable three-unit polyethylene glycol (PEG) linker bearing a terminal primary amine and a folate-targeting moiety. It is designed for use as a spacer in the synthesis of antibody-drug conjugates (ADCs) and other bioconjugates, facilitating folate receptor-mediated targeting in research applications.
  • Cleavable three-unit PEG spacer to separate payload and targeting ligand.
  • Terminal primary amine for straightforward conjugation chemistries, such as NHS esters and activated carboxyls.
  • High purity suitable for research (LCMS reported 99.69%).
  • Water-soluble and compatible with common bioconjugation workflows.
  • Supplied in small research quantities for experimental use.
Catalog No. 50-002-03720