Learn More
Medchemexpress LLC 3,6-Di-tert-butyl-9H-carbazole-d25 | 2569012-64-0 | >98% | 304.57 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW011565S1
Encompass_Preferred
3,6-Di-tert-butyl-9H-carbazole-d25 is a deuterium-labeled version of 3,6-Di-tert-butyl-9H-carbazole. This compound can be utilized as a tracer in various analytical applications and also serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including deuterium, are incorporated into drug molecules primarily for quantitative tracing during the drug development process. Deuteration is particularly noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope-labeled compound.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52528