Learn More
Medchemexpress LLC 3,6-dichloro-4-methylpyridazine | 19064-64-3 | MFCD00006465 | 100.0% | 163.00 g/mol | C5H4Cl2N2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01538810G
Encompass_Preferred
3,6-Dichloro-4-methylpyridazine (CAS 19064-64-3) is a chlorinated pyridazine used as an organic reagent in life-science research and chemical synthesis. Offered in multiple pack sizes, it is suitable as a synthetic building block, for biochemical assays, and for analytical applications.
  • High purity: 99.98%.
  • Molecular formula C5H4Cl2N2 and molecular weight 163.00 g/mol.
  • Suitable for medicinal chemistry and biochemical assay development.
  • Available in multiple pack sizes for experimental flexibility.
  • Storage recommendations provided to preserve stability (store at -20°C under nitrogen; in solvent: -80°C, 6 months; -20°C, 1 month).
  • Characterized with CAS registry number 19064-64-3 for unambiguous identification.
Catalog No. 50-003-02528