Learn More
Medchemexpress LLC 3,6-dichloro-4-methylpyridazine | 19064-64-3 | MFCD00006465 | 100.0% | 163.00 g/mol | C5H4Cl2N2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015388
Encompass_Preferred
3,6-dichloro-4-methylpyridazine is a chlorinated pyridazine supplied as a research-grade biochemical reagent and synthetic building block. It is provided as a solid for life-science and organic synthesis applications that require a high-purity chlorinated heterocycle.
  • High purity (99.98%).
  • Chemical formula C5H4Cl2N2 and molecular weight 163.00 g/mol.
  • Solid, off-white to light yellow appearance.
  • Stable when stored at -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Suitable for research use and organic synthesis applications.
  • Available in gram to bulk quantities.
Catalog No. 50-003-03083