Learn More
Medchemexpress LLC 3,6-Dichlorotrimellitic acid | 137071-78-4 | MFCD16294491 | 99.3% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD08285MG
Encompass_Preferred
3,6-Dichlorotrimellitic acid is a crucial precursor utilized in the synthesis of various dichlorinated fluoresceins and rhodamines, including TET and HEX. These derivatives are widely employed in the field of molecular biology for labeling oligonucleotides and in DNA sequencing applications, highlighting its significance in advanced research methodologies.
  • Key precursor for dichlorinated fluoresceins and rhodamines.
  • Widely used for labeling oligos.
  • Essential in DNA sequencing applications.
Catalog No. 50-004-10238