Learn More
Medchemexpress LLC 3,6-diiodo-9H-carbazole | 57103-02-3 | MFCD00450017 | 99.6% | 419.00 | C12H7I2N | 500 G

Supplier: Medchemexpress LLC HYW022224500G
3,6-Diiodo-9H-carbazole is an iodinated carbazole derivative used as a synthetic building block for the design and synthesis of carbazole-based compounds in laboratory research. Intended for research use only; follow the safety data sheet for handling and storage instructions.
- High purity (≈99.6%).
- Molecular formula C12H7I2N.
- Molecular weight 419.00.
- Solid form for easier handling and storage.
- Store at 4°C and protect from light; in solvent: -80°C for 6 months or -20°C for 1 month.
- Suitable as a building block in synthetic chemistry applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.