Learn More
Medchemexpress LLC 3,6-diiodo-9H-carbazole | 57103-02-3 | MFCD00450017 | 99.6% | 419.00 g/mol | C12H7I2N | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW022224
Encompass_Preferred
3,6-diiodo-9H-carbazole is an iodinated carbazole supplied as a solid for use as an intermediate in organic synthesis. It is commonly employed in the design and preparation of carbazole derivatives and in reactions that require activated aryl iodides, including cross-coupling chemistry.
  • High reported purity (99.6%).
  • Solid form, easy to weigh and handle.
  • Suitable for cross-coupling and electrophilic substitution reactions.
  • Well-characterized with available COA and SDS.
  • Available in multiple pack sizes for research and scale-up.
Catalog No. 50-003-03683