Learn More
Medchemexpress LLC 3,6-Methano-8H-1,5,7-trioxacyclopenta[ij]cycloprop[a]azulene-4,8(3H)-dione, hexahydro... | 17617-45-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1494
Encompass_Preferred
Picrotoxinin is a potent convulsant and chloride channel blocker that acts as a noncompetitive GABAA receptor antagonist. It negatively modulates the action of GABA on GABAA receptors, specifically inhibiting the α1β2γ2L GABAA receptor.
  • Potent convulsant
  • Chloride channel blocker
  • Noncompetitive GABAA receptor antagonist
  • Modulates GABA action on GABAA receptors
  • Inhibits α1β2γ2L GABAA receptor
Catalog No. 50-004-13832